Merck
  • A surface wipe sampling and LC-MS/MS method for the simultaneous detection of six antineoplastic drugs commonly handled by healthcare workers.
A surface wipe sampling and LC-MS/MS method for the simultaneous detection of six antineoplastic drugs commonly handled by healthcare workers.
Analytical and bioanalytical chemistry (2015-07-05)
Matthew Jeronimo, Manuel Colombo, George Astrakianakis, Chun-Yip Hon
ABSTRACT
An effective wipe sampling and LC-MS/MS method was developed to simultaneously analyze six commonly administered antineoplastic drugs in stainless steel surface. The analyzed drugs were methotrexate, paclitaxel, cyclophosphamide, 5-fluorouracil, vincristine, and oxaliplatin, a frequently prepared antineoplastic drug that has not been included among any of the published simultaneous detection methods. The established method was used to evaluate the recoveries of antineoplastic drugs on brand new and worn stainless steel surfaces by wiping the plates with a Whatman filter paper wetted with 0.5 mL of water/methanol (20:80) with 0.1% formic acid followed by LC-MS/MS before desorbing the filter with a water/methanol (50:50) solution. A significant decrease in the recovery of all evaluated drugs was found when worn plates were used. Additionally, the inter-personnel variability on drug recoveries during wiping procedures was evaluated. Significantly higher recoveries were achieved by the personnel with more training and experience versus personnel without prior experience. Finally, a laboratory stability test was developed to assess the degradation of the antineoplastic drugs during replicated shipping conditions. With the exception of vincristine sulfate which exhibited a significant (p < 0.05) degradation after 48 h, all evaluated drugs were stable during the first 24-48 h. However, after 144 h, an increase in the degradation of all evaluated drugs was observed, with oxaliplatin and 5-fluorouracil exhibiting the most degradation.
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Biphenyl, ≥99%
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
SAFC
Methotrexate
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methotrexate-D3 solution, 100 μg/mL in methanol with 0.01N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%