Merck
  • Single-use thermoplastic microfluidic burst valves enabling on-chip reagent storage.
Single-use thermoplastic microfluidic burst valves enabling on-chip reagent storage.
Microfluidics and nanofluidics (2015-05-15)
Omid D Rahmanian, Don L DeVoe
ABSTRACT
A simple and reliable method for fabricating single-use normally closed burst valves in thermoplastic microfluidic devices is presented, using a process flow that is readily integrated into established workflows for the fabrication of thermoplastic microfluidics. An experimental study of valve performance reveals the relationships between valve geometry and burst pressure. The technology is demonstrated in a device employing multiple valves engineered to actuate at different inlet pressures that can be generated using integrated screw pumps. On-chip storage and reconstitution of fluorescein salt sealed within defined reagent chambers are demonstrated. By taking advantage of the low gas and water permeability of cyclic olefin copolymer, the robust burst valves allow on-chip hermetic storage of reagents, making the technology well suited for the development of integrated and disposable assays for use at the point of care.
Sigma-Aldrich
Cyclohexane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Cyclohexane, JIS special grade
Sigma-Aldrich
Cyclohexane, suitable for HPLC
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Fluorescein, for fluorescence, free acid