Merck
The actions of neurotensin in rat bladder detrusor contractility.
Scientific reports (2015-06-09)
Xingyou Dong, Xinyu Bai, Jiang Zhao, Liang Wang, Qingqing Wang, Longkun Li
ABSTRACT
This study assessed the expression, distribution and function of neurotensin (NTs) and two main neurotensin receptors (NTSR), NTSR1 and NTSR2 in normal rat urinary bladders. NTs is primarily located in the suburothelium and the interstitium of smooth muscle bundles. The NTSR1 and NTSR2 receptor subtypes are found to co-localize with smooth muscle cells (SMCs). NTs not only can directly act on bladder SMCs to induce intracellular calcium mobilization by activating the phospholipase C/inositol triphosphate (PLC/IP3) pathway, promoting extracellular calcium influx through a non-selective cation channels, but may be also involved in the modulation of the cholinergic system. Nowadays, the selective antimuscarinic drugs (solifenacin) and the selective beta 3-adrenergic agonist (mirabegron) are used as the first-line pharmacotherapy for overactive bladder (OAB), but without satisfactory treatment benefits in some patients. This study provided evidence suggesting that bladder NTs may play an important role in the regulation of micturition. Further research is needed to investigate the effects of NTs on bladder contractility and the underlying mechanism, which might reveal that the administration of NTSR antagonists can potentially relieve the symptoms of OAB by coordination with antimuscarinic pharmacotherapy.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Anti-NTR2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Rhodamine B, for fluorescence
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Anti-NTSR1 antibody produced in rabbit, affinity isolated antibody
SAFC
Sodium chloride solution, 5 M