Merck
  • Polymer monolith microextraction using poly(butyl methacrylate-co-1,6-hexanediol ethoxylate diacrylate) monolithic sorbent for determination of phenylurea herbicides in water samples.
Polymer monolith microextraction using poly(butyl methacrylate-co-1,6-hexanediol ethoxylate diacrylate) monolithic sorbent for determination of phenylurea herbicides in water samples.
Talanta (2015-11-26)
Shu-Ling Lin, Yu-Ru Wu, Ming-Ren Fuh
ABSTRACT
In this study, recently developed 1,6-hexanediol ethoxylate diacrylate (HEDA)-based polymeric monoliths were utilized as sorbents for efficient extraction of phenylurea herbicides (PUHs) from water samples. The HEDA-based monolithic sorbents were prepared in a fused silica capillary (0.7mm i.d., 4.5-cm long) for polymer monolith microextraction (PMME). The experimental parameters of PMME microextraction including sample loading speed, pH of sample solution, composition of elution solvent, and addition of salt were optimized to efficiently extract PUHs from environmental water samples. The extracted PUHs were determined using ultra-high performance liquid chromatography (UHPLC) with UV-photodiode array detection. The extraction recoveries for PUHs-spiked water samples were 91.1-108.1% with relative standard deviations lower than 5%. The linearity range was 0.025-25ngmL(-1) for each PUH and the detection limits of PUHs were estimated at 0.006-0.019ng mL(-1). In addition, good intra-day/inter-day precision (0.1-8.7%/0.2-8.9%) and accuracy (92.0-108.0%/96.5-105.2%) of the proposed method were obtained. The extraction capacity of the monolith-filled capillary was also determined to be approximately 1μg. Moreover, each monolith-filled capillary could be reused up to 8 times without carry-over. According to the European Union regulations, the allowed permissible limit of any single herbicide in drinking water is 0.1ng mL(-1). This permissible level fell in the linear range examined in this study. In addition, the proposed method provided detection limits lower than the allowed permissible level, which demonstrated the feasibility of utilizing the HEDA-based monolithic sorbent to perform PMME for determining contaminants, such as PUHs, in environmental application.
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Sigma-Aldrich
Lauryl methacrylate, contains 500 ppm MEHQ as inhibitor, 96%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
N-Phenylurea, 97%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Stearyl methacrylate, Mixture of stearyl and cetyl methacrylates, contains MEHQ as inhibitor
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%