Merck
  • Relationship between glycaemic variability and hyperglycaemic clamp-derived functional variables in (impending) type 1 diabetes.
Relationship between glycaemic variability and hyperglycaemic clamp-derived functional variables in (impending) type 1 diabetes.
Diabetologia (2015-09-28)
Annelien Van Dalem, Simke Demeester, Eric V Balti, Katelijn Decochez, Ilse Weets, Evy Vandemeulebroucke, Ursule Van de Velde, An Walgraeve, Nicole Seret, Christophe De Block, Johannes Ruige, Pieter Gillard, Bart Keymeulen, Daniel G Pipeleers, Frans K Gorus
ABSTRACT
We examined whether measures of glycaemic variability (GV), assessed by continuous glucose monitoring (CGM) and self-monitoring of blood glucose (SMBG), can complement or replace measures of beta cell function and insulin action in detecting the progression of preclinical disease to type 1 diabetes. Twenty-two autoantibody-positive (autoAb(+)) first-degree relatives (FDRs) of patients with type 1 diabetes who were themselves at high 5-year risk (50%) for type 1 diabetes underwent CGM, a hyperglycaemic clamp test and OGTT, and were followed for up to 31 months. Clamp variables were used to estimate beta cell function (first-phase [AUC5-10 min] and second-phase [AUC120-150 min] C-peptide release) combined with insulin resistance (glucose disposal rate; M 120-150 min). Age-matched healthy volunteers (n = 20) and individuals with recent-onset type 1 diabetes (n = 9) served as control groups. In autoAb(+) FDRs, M 120-150 min below the 10th percentile (P10) of controls achieved 86% diagnostic efficiency in discriminating between normoglycaemic FDRs and individuals with (impending) dysglycaemia. M 120-150 min outperformed AUC5-10 min and AUC120-150 min C-peptide below P10 of controls, which were only 59-68% effective. Among GV variables, CGM above the reference range was better at detecting (impending) dysglycaemia than elevated SMBG (77-82% vs 73% efficiency). Combined CGM measures were equally efficient as M 120-150 min (86%). Daytime GV variables were inversely correlated with clamp variables, and more strongly with M 120-150 min than with AUC5-10 min or AUC120-150 min C-peptide. CGM-derived GV and the glucose disposal rate, reflecting both insulin secretion and action, outperformed SMBG and first- or second-phase AUC C-peptide in identifying FDRs with (impending) dysglycaemia or diabetes. Our results indicate the feasibility of developing minimally invasive CGM-based criteria for close metabolic monitoring and as outcome measures in trials.
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
Glucose solution, BioUltra, for molecular biology, ~20% in H2O
Sigma-Aldrich
D-(+)-Glucose, SAJ special grade, ≥98.0%
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture