Merck
  • Late-glacial elevated dust deposition linked to westerly wind shifts in southern South America.
Late-glacial elevated dust deposition linked to westerly wind shifts in southern South America.
Scientific reports (2015-07-02)
Heleen Vanneste, François De Vleeschouwer, Antonio Martínez-Cortizas, Clemens von Scheffer, Natalia Piotrowska, Andrea Coronato, Gaël Le Roux
ABSTRACT
Atmospheric dust loadings play a crucial role in the global climate system. Southern South America is a key dust source, however, dust deposition rates remain poorly quantified since the last glacial termination (~17 kyr ago), an important timeframe to anticipate future climate changes. Here we use isotope and element geochemistry in a peat archive from Tierra del Fuego, to reconstruct atmospheric dust fluxes and associated environmental and westerly wind changes for the past 16.2 kyr. Dust depositions were elevated during the Antarctic Cold Reversal (ACR) and second half of the Younger Dryas (YD) stadial, originating from the glacial Beagle Channel valley. This increase was most probably associated with a strengthening of the westerlies during both periods as dust source areas were already available before the onset of the dust peaks and remained present throughout. Congruent with glacier advances across Patagonia, this dust record indicates an overall strengthening of the wind belt during the ACR. On the other hand, we argue that the YD dust peak is linked to strong and poleward shifted westerlies. The close interplay between dust fluxes and climatic changes demonstrates that atmospheric circulation was essential in generating and sustaining present-day interglacial conditions.
Sigma-Aldrich
Gallium, 99.99% trace metals basis
Sigma-Aldrich
Strontium, random pieces, 99%
Sigma-Aldrich
Gallium, 99.999% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Gallium, 99.9995% trace metals basis
Sigma-Aldrich
Cesium, ingot, ≥99.95% trace metals basis
Scandium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Scandium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Scandium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Scandium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Rubidium, ingot, 99.6% trace metals basis
Sigma-Aldrich
Potassium hydride, in paraffin