Merck
  • Anti-cancer fatty-acid derivative induces autophagic cell death through modulation of PKM isoform expression profile mediated by bcr-abl in chronic myeloid leukemia.
Anti-cancer fatty-acid derivative induces autophagic cell death through modulation of PKM isoform expression profile mediated by bcr-abl in chronic myeloid leukemia.
Cancer letters (2015-02-04)
Haruka Shinohara, Kohei Taniguchi, Minami Kumazaki, Nami Yamada, Yuko Ito, Yoshinori Otsuki, Bunji Uno, Fumihiko Hayakawa, Yosuke Minami, Tomoki Naoe, Yukihiro Akao
ABSTRACT
The fusion gene bcr-abl develops chronic myeloid leukemia (CML), and stimulates PI3K/Akt/mTOR signaling, leading to impaired autophagy. PI3K/Akt/mTOR signaling also plays an important role in cell metabolism. The Warburg effect is a well-recognized hallmark of cancer energy metabolism, and is regulated by the mTOR/c-Myc/hnRNP/PKM signaling cascade. To develop a new strategy for the treatment of CML, we investigated the associations among bcr-abl, the cascade related to cancer energy metabolism, and autophagy induced by a fatty-acid derivative that we had previously reported as being an autophagy inducer. Here we report that a fatty-acid derivative, AIC-47, induced transcriptional repression of the bcr-abl gene and modulated the expression profile of PKM isoforms, resulting in autophagic cell death. We show that c-Myc functioned as a transcriptional activator of bcr-abl, and regulated the hnRNP/PKM cascade. AIC-47, acting through the PPARγ/β-catenin pathway, induced down-regulation of c-Myc, leading to the disruption of the bcr-abl/mTOR/hnRNP signaling pathway, and switching of the expression of PKM2 to PKM1. This switching caused autophagic cell death through an increase in the ROS level. Our findings suggest that AIC-47 induced autophagic cell death through the PPARγ/β-catenin/bcr-abl/mTOR/hnRNP/PKM cascade.
Product Description
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Thionyl chloride, ReagentPlus®, 99.5%, low iron
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Thionyl chloride, reagent grade, 97%
Sigma-Aldrich
Thionyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Supelco
5,5-Dimethyl-1-pyrroline N-oxide, for ESR-spectroscopy
Sigma-Aldrich
3-Decenoic acid, AldrichCPR
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%