Merck
  • Combination of aqueous two-phase extraction and cation-exchange chromatography: new strategies for separation and purification of alliin from garlic powder.
Combination of aqueous two-phase extraction and cation-exchange chromatography: new strategies for separation and purification of alliin from garlic powder.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-03-25)
Xiu-min Jiang, Yan-min Lu, Cong-ping Tan, Yan -Liang, Bo Cui
ABSTRACT
In this study, a two-step process combining aqueous two-phase extraction (ATPE) with chromatography was developed for extraction and purification of alliin from garlic powder. The partition coefficient and yield value of alliin in different types of aqueous two-phase system (ATPS) were compared and response surface methodology (RSM) was used for analyzing and optimizing the extraction process. The optimal extraction conditions of 19% (w/w) (NH4)2SO4, 20% (w/w) 1-prpanol, at 30°C, pH 2.35 with 8.54% (w/w) NaCl was chosen based on the higher yield. Compared to the results obtained with the conventional extraction method, this method had an evident advantage on yield (20.4mg/g versus the original yield of 15.0mg/g) and the concentration of alliin in extract solution by ATPE was close to three times of that with conventional extraction. The purification of alliin was carried out with the ammonium form of sulfonic acid cation-exchange resins 001×7. Sample solution with alliin concentration of 1mg/mL was passed through resins and the desorption of alliin was accomplished by water at the flow velocity of 0.5mL/min, 1.5mL/min, respectively. The purity and recovery of alliin after purification were 80% and 76%, respectively.
Sigma-Aldrich
(±)-L-Alliin, ≥90% (HPLC)
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, JIS special grade
Sigma-Aldrich
1-Propanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
1-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Supelco
1-Propanol, analytical standard
Sigma-Aldrich
1-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Propanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, analytical standard
USP
1-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%