Merck
  • Intravenous immunoglobulin treatment in humans suppresses dendritic cell function via stimulation of IL-4 and IL-13 production.
Intravenous immunoglobulin treatment in humans suppresses dendritic cell function via stimulation of IL-4 and IL-13 production.
Journal of immunology (Baltimore, Md. : 1950) (2014-05-09)
Angela S W Tjon, Rogier van Gent, Haziz Jaadar, P Martin van Hagen, Shanta Mancham, Luc J W van der Laan, Peter A W te Boekhorst, Herold J Metselaar, Jaap Kwekkeboom
ABSTRACT
High-dose i.v. Ig (IVIg) is a prominent immunomodulatory therapy for various autoimmune and inflammatory diseases. Recent mice studies suggest that IVIg inhibits myeloid cell function by inducing a cascade of IL-33-Th2 cytokine production causing upregulation of the inhibitory FcγRIIb, as well as by modulating IFN-γ signaling. The purpose of our study was to explore whether and how these mechanisms are operational in IVIg-treated patients. We show that IVIg in patients results in increases in plasma levels of IL-33, IL-4, and IL-13 and that increments in IL-33 levels correlate with rises in plasma IL-4 and IL-13 levels. Strikingly, no upregulation of FcγRIIb expression was found, but instead a decreased expression of the activating FcγRIIa on circulating myeloid dendritic cells (mDCs) after high-dose, but not after low-dose, IVIg treatment. In addition, expression of the signaling IFN-γR2 subunit of the IFN-γR on mDCs was downregulated upon high-dose IVIg therapy. In vitro experiments suggest that the modulation of FcγRs and IFN-γR2 on mDCs is mediated by IL-4 and IL-13, which functionally suppress the responsiveness of mDCs to immune complexes or IFN-γ. Human lymph nodes and macrophages were identified as potential sources of IL-33 during IVIg treatment. Interestingly, stimulation of IL-33 production in human macrophages by IVIg was not mediated by dendritic cell-specific intercellular adhesion molecule-3-grabbing nonintegrin (DC-SIGN). In conclusion, high-dose IVIg treatment inhibits inflammatory responsiveness of mDCs in humans by Th2 cytokine-mediated downregulation of FcγRIIa and IFN-γR2 and not by upregulation of FcγRIIb. Our results suggest that this cascade is initiated by stimulation of IL-33 production that seems DC-SIGN independent.
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)