Merck
Controlled release of fragrant molecules with visible light.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-10-07)
Zhuozhi Wang, Valentine K Johns, Yi Liao
ABSTRACT
Controlled release of odorous molecules is the key to digital scent technology which will add another dimension to electronics. Photorelease is a cold mechanism that promises better temporal and spatial control than thermal release. Herein we report a novel material composed of an acid-sensitive polymer carrying a fragrant aldehyde and a reversible metastable-state photoacid. It releases the fragrant molecule under visible light, and stops releasing it after the light is turned off. A metastable-state photoacid with a fast reverse-reaction rate was developed to quickly stop the release after irradiation. Both the carrier polymer and the photoacid can be reused after all the fragrant molecules have been released. The material combines the advantages of visible-light activity, fast on/off rate, easy preparation, and recyclability, and thus is promising for digital scent technology.
Product Description
Supelco
p-Anisaldehyde, analytical standard
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG
Sigma-Aldrich
(±)-3-Chloro-1,2-propanediol, 98%
Sigma-Aldrich
p-Anisaldehyde, 98%
Sigma-Aldrich
p-Anisaldehyde, natural, FG
Supelco
(±)-3-Chloro-1,2-propanediol, analytical standard