Merck
  • Highly sensitive SERS-based immunoassay of aflatoxin B1 using silica-encapsulated hollow gold nanoparticles.
Highly sensitive SERS-based immunoassay of aflatoxin B1 using silica-encapsulated hollow gold nanoparticles.
Journal of hazardous materials (2014-12-03)
Juhui Ko, Chankil Lee, Jaebum Choo
ABSTRACT
Aflatoxin B1 (AFB1) is a well-known carcinogenic contaminant in foods. It is classified as an extremely hazardous compound because of its potential toxicity to the human nervous system. AFB1 has also been extensively used as a biochemical marker to evaluate the degree of food spoilage. In this study, a novel surface-enhanced Raman scattering (SERS)-based immunoassay platform using silica-encapsulated hollow gold nanoparticles (SEHGNs) and magnetic beads was developed for highly sensitive detection of AFB1. SEHGNs were used as highly stable SERS-encoding nano tags, and magnetic beads were used as supporting substrates for the high-density loading of immunocomplexes. Quantitative analysis of AFB1 was performed by monitoring the intensity change of the characteristic peaks of Raman reporter molecules. The limit of detection (LOD) of AFB1, determined by this SERS-based immunoassay, was determined to be 0.1 ng/mL. This method has some advantages over other analytical methods with respect to rapid analysis (less than 30 min), good selectivity, and reproducibility. The proposed method is expected to be a new analytical tool for the trace analysis of various mycotoxins.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Supelco
Malachite Green chloride, analytical standard
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilane, 95%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Trolamine impurity A, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Ethanolamine, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethanolamine, SAJ first grade, ≥99.0%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, JIS special grade, ≥99.0%
Supelco
Ethanolamine, analytical standard
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, suitable for plant cell culture
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Aucubin, primary reference standard
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%