Merck
  • Optimal structural design of mannosylated nanocarriers for macrophage targeting.
Optimal structural design of mannosylated nanocarriers for macrophage targeting.
Journal of controlled release : official journal of the Controlled Release Society (2014-09-16)
Peiming Chen, Xiaoping Zhang, Lee Jia, Robert K Prud'homme, Zoltan Szekely, Patrick J Sinko
ABSTRACT
Macrophages are involved in a number of diseases, such as HIV infection/AIDS, tuberculosis, tumor development and atherosclerosis. Macrophages possess several cell surface receptors (e.g., the mannose receptor, MR) that may serve as drug delivery cellular portals for nanocarriers (NCs). In this study, the optimal structural configuration for cell uptake of mannosylated poly(ethylene glycol)-conjugate type NCs was determined. A series of NCs were synthesized to systematically evaluate the effects of the number of mannose units (Man), the PEG carrier size and the mPEG spacer length between adjacent mannose units on NC uptake into MR-expressing J774.E murine macrophage-like cells. Among NCs with 0, 1, 2 or 4 units of mannose, the uptake of (Man)2-NC was the highest, suggesting a trade-off between avidity and NC-MR clustering on the cell surface that sterically hinders endocytosis. This optimal (Man)2-NC configuration was built into subsequent NCs to optimize the other two parameters, PEG carrier size and spacer length. NCs with 0, 5, 12, 20, 30 or 40 kDa linear PEG carriers showed an inverse relationship between PEG size and uptake. The 12 kDa PEG carrier was chosen for investigating the third parameter, the Man-Man distance, since it may represent the best trade off (i.e., tissue penetration vs. systemic clearance) for in vivo macrophage targeting. Three (Man)2-PEG12kDa NCs with different Man-Man distances (39, 56 or 89Å) were synthesized. The uptake of the NC with the 56Å distance between mannoses was four- and two-fold higher than NCs with 39Å and 89Å distances, respectively. Confocal microscopy confirmed that the optimized (Man)2-PEG12kDa NC with the 56Å Man-Man distance was internalized via endocytosis consistent with temperature-dependent active uptake. In conclusion, the optimal NC structural parameters for targeting the MR on macrophage-like J774.E cells are (i) a small PEG polymer carrier, (ii) two mannose units per NC and (iii) a 56Å distance between adjacent mannose units.
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Sigma-Aldrich
Fmoc-Ser-OH, ≥97.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Fmoc-Cys(Trt)-OH, ≥95.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Boron trifluoride diethyl etherate, purified by redistillation, ≥46.5% BF3 basis
Sigma-Aldrich
Boron trifluoride diethyl etherate, for synthesis
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade