Merck
  • Protein adsorption on electrospun zinc doped hydroxyapatite containing nylon 6 membrane: kinetics and isotherm.
Protein adsorption on electrospun zinc doped hydroxyapatite containing nylon 6 membrane: kinetics and isotherm.
Journal of colloid and interface science (2014-12-30)
Hamid Esfahani, Molamma P Prabhakaran, Esmaeil Salahi, Ali Tayebifard, Mansour Keyanpour-Rad, Mohamad Reza Rahimipour, Seeram Ramakrishna
ABSTRACT
Surface modification of electrospun polymeric membrane surfaces is a critical step towards the separation process including protein adsorption. In this study, the electrospun Nylon fibers was incorporated with positively charged zinc doped hydroxyapatite (HAp) nanoparticles to study the adsorption of negatively charged proteins, namely bovine serum albumin (BSA). Effects of zinc amount within the atomic structure of HAp (nZH; n=0, 4, 8 At.%) was evaluated on produced scaffolds and consequently protein adsorption. The results showed that the ability of Nylon membrane to adsorb BSA increased with incorporation of nZH nanoparticles within the nylon structure. This phenomenon is appeared to be relate to different electrostatic charge and not to physical characteristic of scaffolds. The incorporated membrane (N-4ZH) by nanoparticles with highest zeta (ξ) potential adsorbed the maximum amount of protein. The adsorption of BSA was best fitted with pseudo-second order kinetic model. The experimental isotherm data were further analyzed by using Langmuir and Freundlich equations. By comparing the correlation coefficients obtained for each linear transformation of isotherm analysis, it was found that the Langmuir equation was the best fit equilibrium model that described the adsorption of BSA on these membranes.
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, reagent grade, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Calcium acetate monohydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Calcium phosphate tribasic, suitable for plant cell culture, BioReagent, powder
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Ammonium phosphate dibasic, JIS special grade, ≥99.0%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Ammonium phosphate dibasic, SAJ first grade, 98.5-101.0%
Sigma-Aldrich
Calcium acetate hydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium acetate hydrate, SAJ first grade, 98.0-102.0%
Sigma-Aldrich
Ammonium phosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Calcium acetate hydrate, ReagentPlus®, ≥99% (titration), powder
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ammonium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Ammonium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Calcium acetate hydrate, BioUltra, ≥99.0% (calc. on dried material, KT)