Merck
  • Lentivirus-mediated RNAi knockdown of the gap junction protein, Cx43, attenuates the development of vascular restenosis following balloon injury.
Lentivirus-mediated RNAi knockdown of the gap junction protein, Cx43, attenuates the development of vascular restenosis following balloon injury.
International journal of molecular medicine (2015-01-28)
Xiao-Jian Han, Min Chen, Tao Hong, Ling-Yu Zhu, Dan He, Jiu-Geng Feng, Li-Ping Jiang
ABSTRACT
Percutaneous coronary intervention [PCI or percutaneous transluminal coronary angioplasty (PTCA)] has been developed into a mature interventional treatment for atherosclerotic cardiovascular disease. However, the long-term therapeutic effect is compromised by the high incidence of vascular restenosis following angioplasty, and the underlying mechanisms of vascular restenosis have not yet been fully elucidated. In the present study, we investigated the role of the gap junction (GJ) protein, connexin 43 (Cx43), in the development of vascular restenosis. To establish vascular restenosis, rat carotid arteries were subjected to balloon angioplasty injury. At 0, 7, 14 and 2 days following balloon injury, the arteries were removed, and the intimal/medial area of the vessels was measured to evaluate the degree of restenosis. We found that the intimal area gradually increased following balloon injury. Intimal hyperplasia and restenosis were particularly evident at 14 and 28 days after injury. In addition, the mRNA and protein expression of Cx43 was temporarily decreased at 7 days, and subsequently increased at 14 and 28 days following balloon injury, as shown by RT-PCR and western blot analysis. To determine the involvement of Cx43 in vascular restenosis, the lentivirus vector expressing shRNA targeting Cx43, Cx43-RNAi-LV, was used to silence Cx43 in the rat carotid arteries. The knockdown of Cx43 effectively attenuated the development of intimal hyperplasia and vascular restenosis following balloon injury. Thus, our data indicate the vital role of the GJ protein, Cx43, in the development of vascular restenosis, and provide new insight into the pathogenesis of vascular restenosis. Cx43 may prove to be a novel potential pharmacological target for the prevention of vascular restenosis following PCI.
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Penicillin G sodium salt, 96.0-102.0%
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse