Merck
  • Low-dose prazosin in combination with 5-HT6 antagonist PRX-07034 has antipsychotic effects.
Low-dose prazosin in combination with 5-HT6 antagonist PRX-07034 has antipsychotic effects.
Canadian journal of physiology and pharmacology (2014-11-28)
Renny Abraham, Ramakrishna Nirogi, Anil Shinde, Shantaveer Irupannanavar
ABSTRACT
An extensive amount of research has focused on the development of new pharmacological agents to treat schizophrenia. Varying from person to person, schizophrenia is a heterogeneous disease with symptoms of positive, negative, and cognitive deficits. PRX-07034, a 5-hydroxytryptamine6 (5-HT6) receptor antagonist has been evaluated for its potential in treating obesity and cognitive deficits. This study evaluated PRX-07034 (0.1, 0.3, and 1.0 mg/kg body mass, by intraperitoneal (i.p.) injection), in combination with a low dose of prazosin (0.3 mg/kg, i.p.), for its antipsychotic potential. The research utilized a stereotypy assay, an open field test, an object recognition task, and prepulse inhibition. Dizocilpine, a non-competitive N-methyl-d-aspartate (NMDA) antagonist, was also administered in the above-mentioned assays as a psychomimetic. The combination of PRX-07034 and prazosin alleviated stereotypy and hyperlocomotor activity while enhancing memory in an object recognition task, and reversed sensory-gating deficits induced by dizocilpine. Examination of the medial prefrontal cortex revealed that a combination of PRX-07034 and prazosin reduced the dizocilpine-mediated increase of 5-HT. These results suggest that the combination of a 5-HT6 antagonist with low doses of prazosin could have therapeutic potential in the treatment of schizophrenia.
Sigma-Aldrich
Prazosin hydrochloride, ≥99.0% (HPLC)
USP
Prazosin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Prazosin hydrochloride, European Pharmacopoeia (EP) Reference Standard