Merck
  • Prilling as manufacturing technique for multiparticulate lipid/PEG fixed-dose combinations.
Prilling as manufacturing technique for multiparticulate lipid/PEG fixed-dose combinations.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-07-11)
A Vervaeck, T Monteyne, L Saerens, T De Beer, J P Remon, C Vervaet
ABSTRACT
This study focused on the evaluation of prilling as a technique for the manufacturing of multiparticulate dosage forms. Prills, providing controlled and immediate drug release, were processed and finally combined in capsules yielding a fixed-dose combination. Metoprolol tartrate (MPT) and hydrochlorothiazide (HCT) were used as controlled and immediate release model drugs, respectively. These drugs were embedded in matrices composed of fatty acids and polyethylene glycol (PEG). In order to tailor drug release from the prills, the type of fatty acid, the PEG molecular weight and the fatty acid/PEG ratio were varied. To provide controlled drug release, MPT was embedded in matrices containing PEG and behenic acid. Using different PEG molecular weights (PEG 4000, 6000 and 10,000), MPT release could be tailored over a wide range. To obtain immediate release, HCT was incorporated in matrices composed of PEG and stearic acid. Since high amounts (at least 60%) of PEG were needed for acceptable immediate release, HCT release was independent on PEG molecular weight. Solid state characterization revealed that MPT crystallinity was decreased, while HCT was molecularly dispersed throughout the matrix. Drug release of both MPT and HCT prills was stable during storage. Compared to a fixed-dose reference, oral co-administration of the MPT and HCT prills to dogs yielded a similar bioavailability for the HCT prills, while the MPT prills resulted in a significant higher bioavailability.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Potassium fluoride, BioUltra, ≥99.5% (F)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, SAJ special grade
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrochlorothiazide, Pharmaceutical Secondary Standard; Certified Reference Material