Merck
  • The immunobiology of cobalt: demonstration of a potential aetiology for inflammatory pseudotumours after metal-on-metal replacement of the hip.
The immunobiology of cobalt: demonstration of a potential aetiology for inflammatory pseudotumours after metal-on-metal replacement of the hip.
The bone & joint journal (2014-09-04)
H Lawrence, D Deehan, J Holland, J Kirby, A Tyson-Capper
ABSTRACT
Abnormal wear of cobalt-containing metal-on-metal joints is associated with inflammatory pseudotumours. Cobalt ions activate human toll-like receptor 4 (TLR4), which normally responds to bacterial lipopolysaccharide (LPS) in sepsis. Activation of TLR4 by LPS increases the expression of chemokines IL-8 and CXCL10, which recruit leukocytes and activated T-cells, respectively. This study was designed to determine whether cobalt induces a similar inflammatory response to LPS by promoting the expression of IL-8 and CXCL10. A human monocytic cell line, derived from acute monocytic leukaemia, was treated with cobalt ions and expression of IL-8 and CXCL10 measured at mRNA and protein levels. Cobalt-treated macrophages showed a 60-fold increase in IL-8 mRNA, and an eightfold increase in production of the mature chemokine (both p < 0.001); expression of the CXCL10 gene and protein was also significantly increased by cobalt (both p < 0.001). Experiments were also performed in the presence of CLI-095, a TLR4-specific antagonist which abrogated the cobalt-mediated increase in IL-8 and CXCL10 expression. These findings suggest that cobalt ions induce inflammation similar to that observed during sepsis by the simultaneous activation of two TLR4-mediated signalling pathways. These pathways result in increased production of IL-8 and CXCL10, and may be implicated in pseudotumour formation following metal-on-metal replacement.
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, reagent grade
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, ACS reagent, 98%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., 98-102%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, suitable for plant cell culture
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture