Merck
  • Three-dimensional organic microlasers with low lasing thresholds fabricated by multiphoton and UV lithography.
Three-dimensional organic microlasers with low lasing thresholds fabricated by multiphoton and UV lithography.
Optics express (2014-06-13)
Vincent W Chen, Nina Sobeshchuk, Clément Lafargue, Eric S Mansfield, Jeannie Yom, Lucas R Johnstone, Joel M Hales, Stefan Bittner, Séverin Charpignon, David Ulbricht, Joseph Lautru, Igor Denisyuk, Joseph Zyss, Joseph W Perry, Melanie Lebental
ABSTRACT
Cuboid-shaped organic microcavities containing a pyrromethene laser dye and supported upon a photonic crystal have been investigated as an approach to reducing the lasing threshold of the cavities. Multiphoton lithography facilitated fabrication of the cuboid cavities directly on the substrate or on the decoupling structure, while similar structures were fabricated on the substrate by UV lithography for comparison. Significant reduction of the lasing threshold by a factor of ~30 has been observed for cavities supported by the photonic crystal relative to those fabricated on the substrate. The lasing mode spectra of the cuboid microresonators provide strong evidence showing that the lasing modes are localized in the horizontal plane, with the shape of an inscribed diamond.
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Supelco
Methyl Isobutyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%, FCC
Supelco
4-Methyl-2-pentanone, analytical standard
Sigma-Aldrich
4-Methyl-2-pentanone, SAJ first grade, ≥99.0%
Sigma-Aldrich
4-Methyl-2-pentanone, JIS special grade, ≥99.5%
Sigma-Aldrich
4-Methyl-2-pentanone, suitable for HPLC, ≥99.5%
Sigma-Aldrich
4-Methyl-2-pentanone, ACS reagent, ≥98.5%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%