Merck
Ascorbic acid transport in brain microvascular pericytes.
Biochemical and biophysical research communications (2015-02-04)
William H Parker, Zhi-Chao Qu, James M May
ABSTRACT
Intracellular vitamin C, or ascorbic acid, has been shown to prevent the apoptosis of cultured vascular pericytes under simulated diabetic conditions. We sought to determine the mechanism by which ascorbate is transported into pericytes prior to exerting this protective effect. Measuring intracellular ascorbate, we found that pericytes display a linear uptake over 30 min and an apparent transport Km of 21 μM, both of which are consistent with activity of the Sodium-dependent Vitamin C Transporter 2 (SVCT2). Uptake of both radiolabeled and unlabeled ascorbate was prevented by inhibiting SVCT2 activity, but not by inhibiting the activity of GLUT-type glucose transporters, which import dehydroascorbate to also generate intracellular ascorbate. Likewise, uptake of dehydroascorbate was prevented with the inhibition of GLUTs, but not by inhibiting the SVCT2, indicating substrate specificity of both transporters. Finally, presence of the SVCT2 in pericytes was confirmed by western blot analysis, and immunocytochemistry was used to localize it to the plasma membrane and intracellular sites. Together, these data clarify previous inconsistencies in the literature, implicate SVCT2 as the pericyte ascorbate transporter, and show that pericytes are capable of concentrating intracellular ascorbate against a gradient in an energy- and sodium-dependent fashion.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
1-(2-Hydroxyethyl)piperazine, 98%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
RIPA Buffer
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard