Merck
  • Time of administration important? Morning versus evening dosing of valsartan.
Time of administration important? Morning versus evening dosing of valsartan.
Journal of hypertension (2014-09-27)
Dion H Zappe, Nora Crikelair, Albert Kandra, Paolo Palatini
ABSTRACT
Studies suggest that bedtime dosing of an angiotensin-converting enzyme (ACE)-inhibitor or angiotensin receptor blocker shows a more sustained and consistent 24-h antihypertensive profile, including greater night-time blood pressure (BP) reduction. We compared the antihypertensive effects of morning (a.m.) and evening (p.m.) dosing of valsartan on 24-h BP. This 26-week, multicentre, randomized, double-blind study evaluated the efficacy and safety of valsartan 320 mg, dosed a.m. or p.m., versus lisinopril 40 mg (a.m.), a long-acting ACE-inhibitor, in patients with grade 1-2 hypertension and at least one additional cardiovascular risk factor. Patients (n = 1093; BP = 156 ± 11/91 ± 8 mmHg; 62 years, 56% male, 99% white) received (1 : 1 : 1) valsartan 160 mg a.m. or p.m. or lisinopril 20 mg a.m. for 4 weeks, then force-titrated to double the initial dose for 8 weeks. At Week 12, hydrochlorothiazide (HCTZ) 12.5 mg was added for 14 weeks if office BP was more than 140/90 mmHg and/or ambulatory BP more than 130/80 mmHg. Mean 24-h ambulatory SBP change from baseline to Weeks 12 and 26 was comparable between valsartan a.m. (-10.6 and -13.3 mmHg) and p.m. (-9.8 and -12.3 mmHg) and lisinopril (-10.7 and -13.7 mmHg). There was no benefit of valsartan p.m. versus a.m. on night-time BP, early morning BP and morning BP surge. Evening dosing also did not improve BP lowering in patients requiring add-on HCTZ or in nondippers at baseline. All treatments were well tolerated. Once-daily dosing of valsartan 320 mg results in equally effective 24-h BP efficacy, regardless of dosing time. ClinicalTrials.gov Identifier: NCT00241124.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, SAJ first grade, ≥98.0
Sigma-Aldrich
Lisinopril, ≥98% (HPLC)
Supelco
Lisinopril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
USP
Lisinopril, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Dibutyl phthalate, SAJ special grade, ≥98.0%
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard