Merck
  • Use of VEGFR-2 targeted ultrasound contrast agent for the early evaluation of response to sorafenib in a mouse model of hepatocellular carcinoma.
Use of VEGFR-2 targeted ultrasound contrast agent for the early evaluation of response to sorafenib in a mouse model of hepatocellular carcinoma.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2014-08-02)
Marco Baron Toaldo, Veronica Salvatore, Sara Marinelli, Chiara Palamà, Maddalena Milazzo, Luca Croci, Laura Venerandi, Mario Cipone, Luigi Bolondi, Fabio Piscaglia
ABSTRACT
The aim of this study was to assess the early response to sorafenib using ultrasound molecular imaging in a murine model of hepatocellular carcinoma (HCC). A xenograft model of HCC was established. Then, mice were divided in two groups and received treatment (sorafenib) or placebo for 14 days. The treatment group was further divided into non-responders and responders according to the degree of growth. Contrast enhanced ultrasound (CEUS) was performed using VEGFR-2 targeted microbubbles (BR55, Bracco Suisse SA, Geneva, Switzerland). Dedicated software was used to quantify the amount of bound microbubbles in the tumor as a numerical value (differential targeted enhancement (dTE)). Tumors were then excised and western blot analysis performed. The dTE values decreased from day 0 to day +14 both in the treatment and control groups, but were lower in the former. The non-responder group had higher dTE levels at day 2 compared to responders (p = 0.019). BR55 appears to be useful in the prediction of response to sorafenib in a xenograft model of HCC.
Product Description
Supelco
Tetracaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Tetracaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Tetracaine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Tetracaine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Tetracaine hydrochloride, ≥99%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%