Merck
  • Oral co-administration of elacridar and ritonavir enhances plasma levels of oral paclitaxel and docetaxel without affecting relative brain accumulation.
Oral co-administration of elacridar and ritonavir enhances plasma levels of oral paclitaxel and docetaxel without affecting relative brain accumulation.
British journal of cancer (2014-05-02)
J J M A Hendrikx, J S Lagas, E Wagenaar, H Rosing, J H M Schellens, J H Beijnen, A H Schinkel
ABSTRACT
The intestinal uptake of the taxanes paclitaxel and docetaxel is seriously hampered by drug efflux through P-glycoprotein (P-gp) and drug metabolism via cytochrome P450 (CYP) 3A. The resulting low oral bioavailability can be boosted by co-administration of P-gp or CYP3A4 inhibitors. Paclitaxel or docetaxel (10 mg/kg) was administered to CYP3A4-humanised mice after administration of the P-gp inhibitor elacridar (25 mg kg(-1)) and the CYP3A inhibitor ritonavir (12.5 mg kg(-1)). Plasma and brain concentrations of the taxanes were measured. Oral co-administration of the taxanes with elacridar increased plasma concentrations of paclitaxel (10.7-fold, P<0.001) and docetaxel (four-fold, P<0.001). Co-administration with ritonavir resulted in 2.5-fold (paclitaxel, P<0.001) and 7.3-fold (docetaxel, P<0.001) increases in plasma concentrations. Co-administration with both inhibitors simultaneously resulted in further increased plasma concentrations of paclitaxel (31.9-fold, P<0.001) and docetaxel (37.4-fold, P<0.001). Although boosting of orally applied taxanes with elacridar and ritonavir potentially increases brain accumulation of taxanes, we found that only brain concentrations, but not brain-to-plasma ratios, were increased after co-administration with both inhibitors. The oral availability of taxanes can be enhanced by co-administration with oral elacridar and ritonavir, without increasing the brain penetration of the taxanes.
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Ritonavir, European Pharmacopoeia (EP) Reference Standard
Anhydrous Docetaxel, European Pharmacopoeia (EP) Reference Standard
USP
Docetaxel, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Elacridar, ≥98% (HPLC)
Sigma-Aldrich
Ritonavir, ≥98% (HPLC)