Merck
  • Acrylonitrile exposure in the general population following a major train accident in Belgium: a human biomonitoring study.
Acrylonitrile exposure in the general population following a major train accident in Belgium: a human biomonitoring study.
Toxicology letters (2014-09-17)
T De Smedt, K De Cremer, C Vleminckx, S Fierens, B Mertens, I Van Overmeire, M Bader, P De Paepe, T Göen, B Nemery, T Schettgen, C Stove, H Van Oyen, J Van Loco, A Van Nieuwenhuyse
ABSTRACT
On Saturday May 4, 2013, a train transporting chemicals derailed in the village of Wetteren (Belgium) and caused a leak of acrylonitrile (ACN). To assess the human exposure to acrylonitrile in the local population with the highest suspected exposure. Between May 18-25, 242 residents participated in the study. N-2-cyanoethylvaline (CEV), a biomarker that is highly specific for ACN exposure, was measured in the blood. To account for potential influence by smoking, cotinine was determined in the urine. Participants also filled in a short questionnaire. In the evacuated zone, 37.3% of the non-smokers and 40.0% of the smokers had CEV concentrations above the reference values of 10 and 200 pmol/g globin, respectively, at the time of the train accident. Spatial mapping of the CEV concentrations depending on the residential address showed a distribution pattern following the sewage system. The train derailment resulted in a highly atypical sequence-of-events. In addition to exposure in the direct vicinity of the site of the train derailment, exposure also occurred via the sewage system, into which acrylonitrile had entered shortly after the accident.
Product Description
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Supelco
Acrylonitrile solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
SAFC
L-Valine
Valine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Supelco
Acrylonitrile, analytical standard
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Acrylonitrile, ≥99%, contains 35-45 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis