Merck
  • Repeatability and reproducibility of retention data and band profiles on reversed-phase liquid chromatography columns. III. Results obtained with Kromasil C18 columns.
Repeatability and reproducibility of retention data and band profiles on reversed-phase liquid chromatography columns. III. Results obtained with Kromasil C18 columns.
Journal of chromatography. A (1999-10-16)
M Kele, G Guiochon
ABSTRACT
The reproducibility of the retention data and the band profiles was investigated with Kromasil C18 columns (silica-based monomeric type reversed-phase packing material). High precision data were obtained and statistically compared among five columns from the same batch (column-to-column reproducibility) and six columns, one from each of six different batches (batch-to-batch reproducibility). These data were acquired under five different sets of chromatographic conditions, for a group of 30 neutral, acidic and basic compounds selected as probes following an experimental protocol previously described. Data characterizing the retention time, the retention factor, the separation factor, the column efficiency and the peak asymmetry for the different probe compounds are reported. Factors describing the silica surface interaction with the selected probe compounds, such as the hydrophobic interaction selectivity, the steric selectivity, and the separation factors of basic compounds at different pH values were also determined. The influence of the underlying silica on these data and correlations between the chromatographic and physico-chemical properties of the different batches are discussed.
Product Description
Kromasil® C18 HPLC Column, 5 μm particle size, pore size 100 Å, L × I.D. 150 mm × 4.6 mm