Merck
  • Genomic analysis of Chthonomonas calidirosea, the first sequenced isolate of the phylum Armatimonadetes.
Genomic analysis of Chthonomonas calidirosea, the first sequenced isolate of the phylum Armatimonadetes.
The ISME journal (2014-01-31)
Kevin C-Y Lee, Xochitl C Morgan, Peter F Dunfield, Ivica Tamas, Ian R McDonald, Matthew B Stott
ABSTRACT
Most of the lineages of bacteria have remained unknown beyond environmental surveys using molecular markers. Until the recent characterisation of several strains, the phylum Armatimonadetes (formerly known as 'candidate division OP10') was a dominant and globally-distributed lineage within this 'uncultured majority'. Here we report the first Armatimonadetes genome from the thermophile Chthonomonas calidirosea T49(T) and its role as a saccharide scavenger in a geothermal steam-affected soil environment. Phylogenomic analysis indicates T49(T) to be related closely to the phylum Chloroflexi. The predicted genes encoding for carbohydrate transporters (27 carbohydrate ATP-binding cassette transporter-related genes) and carbohydrate-metabolising enzymes (including at least 55 putative enzymes with glycosyl hydrolase domains) within the 3.43 Mb genome help explain its ability to utilise a wide range of carbohydrates as well as its inability to break down extracellular cellulose. The presence of only a single class of branched amino acid transporter appears to be the causative step for the requirement of isoleucine for growth. The genome lacks many commonly conserved operons (for example, lac and trp). Potential causes for this, such as dispersion of functionally related genes via horizontal gene transfer from distant taxa or recent genome recombination, were rejected. Evidence suggests T49(T) relies on the relatively abundant σ-factors, instead of operonic organisation, as the primary means of transcriptional regulation. Examination of the genome with physiological data and environmental dynamics (including interspecific interactions) reveals ecological factors behind the apparent elusiveness of T49(T) to cultivation and, by extension, the remaining 'uncultured majority' that have so far evaded conventional microbiological techniques.
Product Description
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, acid washed, powder, for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm