Merck
  • The genotoxicity of sodium saccharin and sodium chloride in relation to their cancer-promoting properties.
The genotoxicity of sodium saccharin and sodium chloride in relation to their cancer-promoting properties.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1985-04-01)
J Ashby
ABSTRACT
The literature indicates that sodium saccharin is non-reactive to DNA and inactive as a gene mutagen in vitro. At elevated dose levels it is capable of producing structural disturbances in eukaryotic chromosomes in vitro, and it shows intermittent activity as a very weak germ-cell and somatic-cell mutagen in vivo. Its possible mode of action in these respects is speculated on and related to its ability to promote bladder tumours in rats at elevated dose levels. A review of the toxicology of sodium chloride reveals a profile of genotoxic activities almost identical to that of sodium saccharin. It is suggested that the recorded genotoxic and cancer-promoting activities of these chemicals will only become apparent at elevated dose levels that define them as significant contributors to the biological medium (solvent) rather than as trace xenobiotic toxins (solutes). The possible activity of acid saccharin, or of its potassium, calcium and ammonium salts, as ionic genotoxins requires urgent evaluation.
Sigma-Aldrich
Sodium standard solution, suitable for atomic absorption spectrometry, 1000 ppm Na, 1 mg/mL Na
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Saccharin, European Pharmacopoeia (EP) Reference Standard
Supelco
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Saccharin, ≥99%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Saccharin, ≥98%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
USP
Saccharin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloride standard solution, 1 mg/mL Cl-