Merck
  • Engineering dextran-based scaffolds for drug delivery and tissue repair.
Engineering dextran-based scaffolds for drug delivery and tissue repair.
Nanomedicine (London, England) (2012-12-06)
Guoming Sun, Jeremy J Mao
ABSTRACT
Owing to its chemically reactive hydroxyl groups, dextran can be modified with different functional groups to form spherical, tubular and 3D network structures. The development of novel functional scaffolds for efficient controlled release and tissue regeneration has been a major research interest, and offers promising therapeutics for many diseases. Dextran-based scaffolds are naturally biodegradable and can serve as bioactive carriers for many protein biomolecules. The reconstruction of the in vitro microenvironment with proper signaling cues for large-scale tissue regenerative scaffolds has yet to be fully developed, and remains a significant challenge in regenerative medicine. This paper will describe recent advances in dextran-based polymers and scaffolds for controlled release and tissue engineering. Special attention is given to the development of dextran-based hydrogels that are precisely manipulated with desired structural properties and encapsulated with defined angiogenic growth factors for therapeutic neovascularization, as well as their potential for wound repair.
Product Description
Supelco
Dextran, analytical standard, for GPC, 150,000
Supelco
Dextran, analytical standard, for GPC, 410,000
Supelco
Dextran, analytical standard, for GPC, 670,000
Supelco
Dextran, analytical standard, for GPC, 12,000
Supelco
Dextran, analytical standard, for GPC, 270,000
Supelco
Dextran, analytical standard, for GPC, 80,000
Supelco
Dextran, analytical standard, for GPC, 50,000
Dextran 4 for calibration, Dextran 4 European Pharmacopoeia (EP) Reference Standard
Dextran Vo, Dextran Vo European Pharmacopoeia (EP) Reference Standard
Dextran 1, Dextran 1 European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
Supelco
Dextran, analytical standard, for GPC, 1,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Supelco
Dextran, analytical standard, for GPC, 5,000