Merck
Nicotinic acid: an old drug with a promising future.
British journal of pharmacology (2007-11-27)
E T Bodor, S Offermanns
ABSTRACT
Nicotinic acid has been used for decades to treat dyslipidaemic states. In particular its ability to raise the plasma HDL cholesterol concentration has led to an increased interest in its pharmacological potential. The clinical use of nicotinic acid is somewhat limited due to several harmless but unpleasant side effects, most notably a cutaneous flushing phenomenon. With the recent discovery of a nicotinic acid receptor, it has become possible to better understand the mechanisms underlying the metabolic and vascular effects of nicotinic acid. Based on these new insights into the action of nicotinic acid, novel strategies are currently under development to maximize the pharmacological potential of this drug. The generation of both flush-reducing co-medications of nicotinic acid and novel drugs targeting the nicotinic acid receptor will provide future therapeutic options for the treatment of dyslipidaemic disorders.
Sigma-Aldrich
Nicotinic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinic acid, European Pharmacopoeia (EP) Reference Standard
USP
Niacin, United States Pharmacopeia (USP) Reference Standard