Merck
  • Pharmacokinetic drug interactions involving 17alpha-ethinylestradiol: a new look at an old drug.
Pharmacokinetic drug interactions involving 17alpha-ethinylestradiol: a new look at an old drug.
Clinical pharmacokinetics (2007-01-27)
Hongjian Zhang, Donghui Cui, Bonnie Wang, Yong-Hae Han, Praveen Balimane, Zheng Yang, Michael Sinz, A David Rodrigues
ABSTRACT
17alpha-Ethinylestradiol (EE) is widely used as the estrogenic component of oral contraceptives (OC). In vitro and in vivo metabolism studies indicate that EE is extensively metabolised, primarily via intestinal sulfation and hepatic oxidation, glucuronidation and sulfation. Cytochrome P450 (CYP)3A4-mediated EE 2-hydroxylation is the major pathway of oxidative metabolism of EE. For some time it has been known that inducers of drug-metabolising enzymes (such as the CYP3A4 inducer rifampicin [rifampin]) can lead to breakthrough bleeding and contraceptive failure. Conversely, inhibitors of drug-metabolising enzymes can give rise to elevated EE plasma concentrations and increased risks of vascular disease and hypertension. In vitro studies have also shown that EE inhibits a number of human CYP enzymes, such as CYP2C19, CYP3A4 and CYP2B6. Consequently, there are numerous reports in the literature describing EE-containing OC formulations as perpetrators of pharmacokinetic drug interactions. Because EE may participate in multiple pharmacokinetic drug interactions as either a victim or perpetrator, pharmaceutical companies routinely conduct clinical drug interaction studies with EE-containing OCs when evaluating new chemical entities in development. It is therefore critical to understand the mechanisms underlying these drug interactions. Such an understanding can enable the interpretation of clinical data and lead to a greater appreciation of the profile of the drug by physicians, clinicians and regulators. This article summarises what is known of the drug-metabolising enzymes and transporters governing the metabolism, disposition and excretion of EE. An effort is made to relate this information to known clinical drug-drug interactions. The inhibition and induction of drug-metabolising enzymes by EE is also reviewed.
Product Description
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
USP
Ethinyl estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
17α-Ethynylestradiol, ≥98%
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethinyl Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Ethinylestradiol for system suitability, European Pharmacopoeia (EP) Reference Standard