Merck
  • Review of pharmacokinetic, pharmacodynamic and clinical studies with a modern combination of amoxicillin/sulbactam.
Review of pharmacokinetic, pharmacodynamic and clinical studies with a modern combination of amoxicillin/sulbactam.
Chemotherapy (2006-05-25)
J Soutric, C Bantar, N Caruso, R Heguilén, J M Casellas, J M Casellas, A Farinati, A Jasovich, H Arenoso, M Rodriguez
ABSTRACT
Amoxicillin/sulbactam is a modern antimicrobial combination. This combination proved to be useful for the treatment of several infections caused by different microorganisms, mainly with the beta-lactamase-producing species. In this review we present the most relevant pharmacokinetic, pharmacodynamic and clinical information associated with its use.
Sigma-Aldrich
Sulbactam
Sulbactam, European Pharmacopoeia (EP) Reference Standard
Sulbactam for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis