Merck
Raman spectroscopy of graphite.
Philosophical transactions. Series A, Mathematical, physical, and engineering sciences (2004-10-16)
Stephanie Reich, Christian Thomsen
ABSTRACT
We present a review of the Raman spectra of graphite from an experimental and theoretical point of view. The disorder-induced Raman bands in this material have been a puzzling Raman problem for almost 30 years. Double-resonant Raman scattering explains their origin as well as the excitation-energy dependence, the overtone spectrum and the difference between Stokes and anti-Stokes scattering. We develop the symmetry-imposed selection rules for double-resonant Raman scattering in graphite and point out misassignments in previously published works. An excellent agreement is found between the graphite phonon dispersion from double-resonant Raman scattering and other experimental methods.
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic