Merck
  • Epsilon-aminocaproic acid and renal complications: case report and review of the literature.
Epsilon-aminocaproic acid and renal complications: case report and review of the literature.
Clinical nephrology (2002-07-27)
G Manjunath, A Fozailoff, D Mitcheson, M J Sarnak
ABSTRACT
Epsilon-aminocaproic acid (EACA) is a potent anti-fibrinolytic agent that is used in the treatment of excessive bleeding resulting from a systemic fibrinolytic state. It can also be used to treat hematuria through its action on decreasing urinary fibrinolysis. A broad range of renal complications has been ascribed to EACA. Although they are rare, they may be life-threatening and should therefore be immediately recognized.
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)
Sigma-Aldrich
6-Aminocaproic acid, BioUltra, ≥99%
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Aminocaproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Aminocaproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
6-Aminocaproic acid, SAJ special grade, ≥99.0%