Merck
  • Gene expression analysis in gonads and brain of catfish Clarias batrachus after the exposure of malathion.
Gene expression analysis in gonads and brain of catfish Clarias batrachus after the exposure of malathion.
Ecotoxicology and environmental safety (2014-01-29)
Y Prathibha, R Murugananthkumar, A Rajakumar, C Laldinsangi, C C Sudhakumari, S K Mamta, A Dutta-Gupta, B Senthilkumaran
ABSTRACT
Pesticides like malathion have the potential to disrupt development and reproduction of aquatic organisms including fishes. To investigate the likely consequences of malathion exposure at low doses in juvenile catfish, Clarias batrachus, we studied the expression pattern of genes encoding certain transcription factors, activin A, sex steroid or orphan nuclear receptors and steroidogenic enzymes which are known to be involved in gonadal development along with histological changes. To compare further, we also analyzed certain brain specific genes related to gonadal axis. Fifty days post hatch catfish fingerlings were exposed continuously to 1 and 10 µg/L of malathion for 21 days. Results from these experiments indicated that transcript levels of various genes were altered by the treatments, which may further affect the gonadal development either directly or indirectly through brain. Histological analysis revealed slow progression of spermatogenesis in testis, while in ovary, the oil droplet oocytes were found to be higher after treatment (10 µg/L). Our findings revealed that the exposure of malathion, even at low doses, hinder or modulate early gonadal development differentially by targeting gene expression pattern of transcription factors, activin A, sex steroid or orphan nuclear receptors and steroidogenic enzymes with an evidence on histological changes. Further, some of the genes showed differential expression at the level of brain in male and female sex after the exposure of malathion.
Product Description
Supelco
Malathion, PESTANAL®, analytical standard
Supelco
Malathion solution, 100 μg/mL in cyclohexane, PESTANAL®, analytical standard
Supelco
Malathion, certified reference material, TraceCERT®
Malathion, European Pharmacopoeia (EP) Reference Standard
USP
Malathion, United States Pharmacopeia (USP) Reference Standard