Merck
  • Reversible anionic redox chemistry in high-capacity layered-oxide electrodes.
Reversible anionic redox chemistry in high-capacity layered-oxide electrodes.
Nature materials (2013-07-16)
M Sathiya, G Rousse, K Ramesha, C P Laisa, H Vezin, M T Sougrati, M-L Doublet, D Foix, D Gonbeau, W Walker, A S Prakash, M Ben Hassine, L Dupont, J-M Tarascon
ABSTRACT
Li-ion batteries have contributed to the commercial success of portable electronics and may soon dominate the electric transportation market provided that major scientific advances including new materials and concepts are developed. Classical positive electrodes for Li-ion technology operate mainly through an insertion-deinsertion redox process involving cationic species. However, this mechanism is insufficient to account for the high capacities exhibited by the new generation of Li-rich (Li(1+x)Ni(y)Co(z)Mn(1-x-y-z)O₂) layered oxides that present unusual Li reactivity. In an attempt to overcome both the inherent composition and the structural complexity of this class of oxides, we have designed structurally related Li₂Ru(1-y)Sn(y)O₃ materials that have a single redox cation and exhibit sustainable reversible capacities as high as 230 mA h g(-1). Moreover, they present good cycling behaviour with no signs of voltage decay and a small irreversible capacity. We also unambiguously show, on the basis of an arsenal of characterization techniques, that the reactivity of these high-capacity materials towards Li entails cumulative cationic (M(n+)→M((n+1)+)) and anionic (O(2-)→O₂(2-)) reversible redox processes, owing to the d-sp hybridization associated with a reductive coupling mechanism. Because Li₂MO₃ is a large family of compounds, this study opens the door to the exploration of a vast number of high-capacity materials.
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium carbonate, battery grade, ≥99.9% trace metals basis