Merck
  • Drug-metabolizing activity of human and rat liver, lung, kidney and intestine slices.
Drug-metabolizing activity of human and rat liver, lung, kidney and intestine slices.
Xenobiotica; the fate of foreign compounds in biological systems (2002-06-18)
R De Kanter, M H De Jager, A L Draaisma, J U Jurva, P Olinga, D K F Meijer, G M M Groothuis
ABSTRACT
1. Organ-specific biotransformation was studied in human and rat liver, lung, kidney and small intestine slices and compared on a protein basis, using four model substances. 2. Deethylation of lidocaine was highest in liver slices from both man and rat, followed by the small intestine. 3. Metabolism of testosterone was highest in liver slices, but a different overall metabolic pattern was found between the different organs. 4. Lung, kidney and intestine slices prepared from human and rat organs showed mainly an unknown metabolite of 7-ethoxycoumarin identified as 4-ethoxy-2-hydroxyphenyl propionic acid (EPPA). 5. The maximal metabolism of 7-ethoxycoumarin in slices was equal with in vivo V(max) in the rat. 6. Phase II metabolism of 7-hydroxycoumarin in kidney and intestinal slices was about 60% of the activity in liver slices. 7. In conclusion, organs other than the liver show a surprisingly high drug-metabolizing activity. Thus, the use of precision-cut slices of a combination of drug metabolizing organs in an in vitro test system from both animal and human origin is required for a proper systematic prediction of drug metabolism in man.
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, ≥99.5%