Merck
  • Increased fructose concentrations in blood and urine in patients with diabetes.
Increased fructose concentrations in blood and urine in patients with diabetes.
Diabetes care (2002-01-30)
Takahiro Kawasaki, Hiroshi Akanuma, Toshikazu Yamanouchi
ABSTRACT
To investigate fructose metabolic changes in patients with diabetes. Serum and urinary fructose concentrations were determined in healthy subjects (n = 23) and in nondiabetic (n = 23) and diabetic patients (n = 26). Fructose was measured using our newly developed method, and (13)C(6)-fructose was used as the internal standard. After adding sample to a fixed amount of internal standard, ion-exchange resins and high-performance liquid chromatography pretreatments were performed. Then, the amount of fructose in the sample was measured by gas chromatography-mass spectrometry. Serum fructose concentrations in patients with diabetes (12.0 +/- 3.8 micromol/l) were significantly higher than those in healthy subjects (8.1 +/- 1.0 micromol/l, P < 0.001) and nondiabetic patients (7.7 +/- 1.6 micromol/l, P < 0.001), and daily urinary fructose excretion was significantly greater in patients with diabetes (127.8 +/- 106.7 micromol/day) than in nondiabetic patients (37.7 +/- 23.0 micromol/day, P < 0.001). In patients with diabetes (n = 20), serum fructose concentrations (8.6 +/- 1.8 micromol/l, P < 0.001) and daily urinary fructose excretion (63.4 +/- 63.8 micromol/day, P < 0.01) significantly decreased by week 2 after admission. The present results differed from those of previous studies in that we found that the serum and urinary fructose concentrations decreased rapidly, concomitant with an improvement in glycemia. Therefore, hyperglycemia was associated with increased serum and urinary fructose concentrations in patients with diabetes.
Sigma-Aldrich
1-Butanol, suitable for HPLC
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Supelco
Acetone, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Supelco
1-Butanol, suitable for HPLC, 99.8%
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%