Merck
Signalling by adhesion receptors.
Nature cell biology (2001-01-09)
S J Fashena, S M Thomas
ABSTRACT
Over the past twenty years, intensive research has enabled us to identify components of specific signalling pathways downstream of an array of adhesion and growth-factor receptors. The first Gordon Research Conference on 'Signalling by Adhesion Receptors', which took place in Newport, Rhode Island, USA (23-28 July 2000), focused on these findings.
Sigma-Aldrich
MAPTRIX-L-YIGSR, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-RKRLQVQLSIRT, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-NRWHSIYITRFG, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-YIGSR, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-NRWHSIYITRFG, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-F-REDV, Fibronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-RQVFQVAYIIIKA, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-TWYKIAFQRNRK, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-KNSFMALYLSKG, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-F-REDV, Fibronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-TWYKIAFQRNRK, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C-TAIPSCPEGTVPLYS, Collagen Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-E-LRAHAVDING, Cadherin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-F-PHSRN, Fibronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-RKRLQVQLSIRT, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-M-VAEIDGIEL, Tenascin-C Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-RQVFQVAYIIIKA, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-KNSFMALYLSKG, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-M-VAEIDGIEL, Tenascin-C Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C-DGEA, Collagen Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-IKVAV, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-F- PHSRN, Fibronectin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C-GLPGER, Collagen Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C-DGEA, Collagen Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-L-IKVAV, Laminin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-E-LFSHAVSSNG, Cadherin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C-GEFYFDLRLKGDK, Collagen Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-E- LFSHAVSSNG, Cadherin Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C-GEFYFDLRLKGDK, Collagen Mimetic, aqueous solution
Sigma-Aldrich
MAPTRIX-C-TAIPSCPEGTVPLYS, Collagen Mimetic, aqueous solution