Merck
Attractive forces between cation condensed DNA double helices.
Biophysical journal (2008-03-11)
Brian A Todd, V Adrian Parsegian, Akira Shirahata, T J Thomas, Donald C Rau
ABSTRACT
By combining single-molecule magnetic tweezers and osmotic stress on DNA assemblies, we separate attractive and repulsive components of the total intermolecular interaction between multivalent cation condensed DNA. Based on measurements of several different cations, we identify two invariant properties of multivalent cation-mediated DNA interactions: repulsive forces decay exponentially with a 2.3 +/- 0.1 A characteristic decay length and the attractive component of the free energy is always 2.3 +/- 0.2 times larger than the repulsive component of the free energy at force-balance equilibrium. These empirical constraints are not consistent with current theories that attribute DNA-DNA attractions to a correlated lattice of counterions. The empirical constraints are consistent with theories for Debye-Hückel interactions between helical line charges and with the order-parameter formalism for hydration forces. Each of these theories posits exponentially decaying attractions and, if we assume this form, our measurements indicate a cation-independent, 4.8 +/- 0.5 A characteristic decay length for intermolecular attractions between condensed DNA molecules.
Sigma-Aldrich
Hexammine cobalt(III) chloride, for use in transformations, X-ray crystallography and NMR
Sigma-Aldrich
Lambda Phage DNA, Methylated from Escherichia coli host strain W3110, solution
Sigma-Aldrich
Spermidine, BioReagent, for molecular biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Deoxynucleotide Set, 100 mM, Individual dNTPs for routine PCR; 0.25 mL each
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Spermidine trihydrochloride, ≥98% (TLC)