Merck
  • Cytofluorometry on cells isolated from paraffin sections after blocking of the background fluoroscence by azocarmine G.
Cytofluorometry on cells isolated from paraffin sections after blocking of the background fluoroscence by azocarmine G.
Histochemistry (1981-01-01)
T Takamatsu, K Nakanishi, M Fukuda, S Fujita
ABSTRACT
A technique for isolation of cells from paraffin embedded tissue is indispensable for the performance of Feulgen-DNA cytofluorometry in parallel with the definition of histological characteristics. Background fluorescence due to nonspecific dye-binding by a "pseudo-plasmal reaction" is usually found to be so intense on cells isolated from formalin-fixed tissues, that we are often forced to abandon quantitative DNA determinations. In the present work, we report the fixation of tissues with Carnoy's fixative for 12 h at 5 degrees C not only reduces nonspecific dye-binding but also facilitates the process of cell isolation. Furthermore, we find that pre-treatment of cells isolated from Carnoy-fixed tissues with acidic azocarmin G solution completely blocks nonspecific dye-binding in subsequent acriflavine Feulgen nuclear staining. This combination of techniques for specimen preparation enables us to carry out Feulgen-DNA cytofluorometry on cells isolated from histological sections with satisfactorily low coefficients of variation (less than 8%). The techniques should be widely applicable for parallel DNA determinations and histology.
Sigma-Aldrich
Azocarmine G