Merck
  • Novel planar and star-shaped molecules: synthesis, electrochemical and photophysical properties.
Novel planar and star-shaped molecules: synthesis, electrochemical and photophysical properties.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-03-15)
Qingfen Niu, Yunqiang Lu, Hongjian Sun, Xiaoyan Li
ABSTRACT
Three novel planar and star-shaped molecules containing thiophene-functionalized group and acetylenic spacers, 4-((5″-iodo-[2,2':5',2″-terthiophen]-5-yl)ethynyl)aniline (I3TEA), 4,4'-([2,2':5',2″-terthiophene]-5,5″-diylbis(ethyne-2,1-diyl))dianiline (3TDDA), 1,3,5-tris((5-((trimethylsilyl)ethynyl)thiophen-2-yl)ethynyl)benzene (TETEB) were synthesized through Sonogashira coupling reaction. Their photophysical and electrochemical properties were explored by UV-vis, photoluminescence (PL) emission and DFT calculation. Three compounds possess unusual phenomena of PL, with the concentration of gradually decreased, the intensity of PL firstly increased and then decreased, this may be due to the aggregate-enhanced emission (AEE) effect and aggregation-caused quenching (ACQ) effect. Their spectroscopic data demonstrate that D-π-A-π-D structured organic dye 3TDDA based on NH2 as donor units and 3T as acceptor units with acetylenic spacers π-conjugated chain between them exhibits good luminescent properties. Dye I3TEA with donor(D)-acceptor(A) structure shows good charge transfer because of amino functional group and iodine atom. Octupolar organic π-conjugated star-shaped molecule TETEB possesses excellent luminescent properties because of its unique structure applied in the optoelectronic field. The redox curves and results of DFT calculations suggest that three compounds have low ionization potential and low Eoxd. All the good properties demonstrate three compounds could be promising candidates for photovoltaic materials.
Sigma-Aldrich
Benzene, JIS special grade, ≥99.5%
Sigma-Aldrich
Benzene, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Benzene, SAJ first grade, ≥99.0%
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material