Merck
  • Mitochondrial complex I activity and NAD+/NADH balance regulate breast cancer progression.
Mitochondrial complex I activity and NAD+/NADH balance regulate breast cancer progression.
The Journal of clinical investigation (2013-02-22)
Antonio F Santidrian, Akemi Matsuno-Yagi, Melissa Ritland, Byoung B Seo, Sarah E LeBoeuf, Laurie J Gay, Takao Yagi, Brunhilde Felding-Habermann
ABSTRACT
Despite advances in clinical therapy, metastasis remains the leading cause of death in breast cancer patients. Mutations in mitochondrial DNA, including those affecting complex I and oxidative phosphorylation, are found in breast tumors and could facilitate metastasis. This study identifies mitochondrial complex I as critical for defining an aggressive phenotype in breast cancer cells. Specific enhancement of mitochondrial complex I activity inhibited tumor growth and metastasis through regulation of the tumor cell NAD+/NADH redox balance, mTORC1 activity, and autophagy. Conversely, nonlethal reduction of NAD+ levels by interfering with nicotinamide phosphoribosyltransferase expression rendered tumor cells more aggressive and increased metastasis. The results translate into a new therapeutic strategy: enhancement of the NAD+/NADH balance through treatment with NAD+ precursors inhibited metastasis in xenograft models, increased animal survival, and strongly interfered with oncogene-driven breast cancer progression in the MMTV-PyMT mouse model. Thus, aberration in mitochondrial complex I NADH dehydrogenase activity can profoundly enhance the aggressiveness of human breast cancer cells, while therapeutic normalization of the NAD+/NADH balance can inhibit metastasis and prevent disease progression.
Sigma-Aldrich
Nicotinic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid sodium salt, 98%
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Supelco
Niacin (Nicotinic Acid), Pharmaceutical Secondary Standard; Certified Reference Material