Merck
  • Phase 2 study of arsenic trioxide followed by autologous hematopoietic cell transplantation for relapsed acute promyelocytic leukemia.
Phase 2 study of arsenic trioxide followed by autologous hematopoietic cell transplantation for relapsed acute promyelocytic leukemia.
Blood (2013-02-16)
Masamitsu Yanada, Motohiro Tsuzuki, Hiroyuki Fujita, Katsumichi Fujimaki, Shin Fujisawa, Kazutaka Sunami, Masafumi Taniwaki, Akira Ohwada, Kosuke Tsuboi, Akio Maeda, Akihiro Takeshita, Shigeki Ohtake, Yasushi Miyazaki, Yoshiko Atsuta, Yukio Kobayashi, Tomoki Naoe, Nobuhiko Emi
ABSTRACT
The optimal treatments for relapsed acute promyelocytic leukemia (APL) remain equivocal. We conducted a phase 2 study to evaluate the efficacy and feasibility of a sequential treatment consisting of induction and consolidation with arsenic trioxide (ATO), peripheral blood stem cell (PBSC) harvest after high-dose cytarabine chemotherapy, and autologous hematopoietic cell transplantation (HCT). Between 2005 and 2009, 35 patients (26 with hematologic and 9 with molecular relapse) were enrolled. Induction therapy resulted in complete remission in 81% of those with hematologic relapse, and most patients became negative for PML-RARα after the first ATO consolidation course, but 4 remained positive. Administration of the second ATO consolidation course further decreased the transcript levels in 3 patients. In total, 25 patients proceeded to PBSC harvest, all of whom successfully achieved the target CD34+ cell doses, and 23 underwent autologous HCT with PML-RARα-negative PBSC graft. Posttransplant relapse occurred in 3 patients, and there was no transplant-related mortality. With a median follow-up of 4.9 years, the 5-year event-free and overall survival rates were 65% and 77%, respectively. These findings demonstrate the outstanding efficacy and feasibility of the sequential treatment featuring ATO and autologous HCT for relapsed APL. This study was registered at http://www.umin.ac.jp/ctr/ as #C000000302.
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis
Supelco
Arsenic(III) oxide, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Arsenic(III) oxide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Arsenic(III) oxide, ACS reagent (primary standard)
Sigma-Aldrich
Arsenic(III) oxide, SAJ first grade, ≥99.0%