Merck
  • cRGD-functionalized polymeric magnetic nanoparticles as a dual-drug delivery system for safe targeted cancer therapy.
cRGD-functionalized polymeric magnetic nanoparticles as a dual-drug delivery system for safe targeted cancer therapy.
Pharmacological research (2013-02-05)
Jian-Min Shen, Fei-Yun Gao, Tao Yin, Hai-Xia Zhang, Ming Ma, Yan-Jie Yang, Feng Yue
ABSTRACT
In this paper we give a method of integrated treatment for cancer and drug-induced complications in the process of cancer therapy through dual-drug delivery system (DDDS). Two hydrophilic drugs, doxorubicin (an antitumor drug) and verapamil (an antiangiocardiopathy drug) combined preliminarily with chitosan shell coated on magnetic nanoparticles (MNPs), followed by entrapping into the PLGA nanoparticles. Further modification was conducted by conjugating tumor-targeting ligand, cyclo(Arg-Gly-Asp-D-Phe-Lys) (c(RGDfK)) peptide, onto the end carboxyl groups on the PLGA-NPs. The size of the resulting cRGD-DOX/VER-MNP-PLGA NPs was approximately 144nm under simulate physiological environment. Under present experiment condition, the entrapment efficiencies of DOX and VER were approximately 74.8 and 53.2wt% for cRGD-DOX/VER-MNP-PLGA NPs. This paper contains interesting pilot data such as NIR-triggered drug release, in vivo drug distribution studies and whole-mouse optical imaging. Histopathological examinations and electrocardiogram comparison demonstrated that the intelligent DDDS could markedly inhibit the growth of tumor and potentially offer an approach for safe cancer therapy.
Product Description
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard