Merck
Reactions of difluorocarbene with organozinc reagents.
Organic letters (2013-02-02)
Vitalij V Levin, Artem A Zemtsov, Marina I Struchkova, Alexander D Dilman
ABSTRACT
Reactions of difluorocarbene with benzyl and alkylzinc halides leading to fluorinated organozinc species have been described. The generated α-difluorinated organozinc reagents are reasonably stable in solution and can be quenched with external electrophiles (iodine, bromine, proton), affording compounds containing the CF(2) fragment.
Sigma-Aldrich
Bromine water, CP
Sigma-Aldrich
Bromine, ≥99.99% trace metals basis
Sigma-Aldrich
Bromine, ACS reagent, ≥99.5%
Sigma-Aldrich
Bromine, reagent grade
Sigma-Aldrich
Bromine, JIS special grade, ≥99.0%
Sigma-Aldrich
Bromine, SAJ first grade, ≥97.0%