Merck
  • Population pharmacokinetics of epsilon-aminocaproic acid in infants undergoing craniofacial reconstruction surgery.
Population pharmacokinetics of epsilon-aminocaproic acid in infants undergoing craniofacial reconstruction surgery.
British journal of anaesthesia (2013-01-29)
P A Stricker, A F Zuppa, J E Fiadjoe, L G Maxwell, E M Sussman, E Y Pruitt, T K Goebel, M R Gastonguay, J A Taylor, S P Bartlett, M S Schreiner
ABSTRACT
Understanding the clinical pharmacology of the antifibrinolytic epsilon-aminocaproic acid (EACA) is necessary for rational drug administration in children. The aim of this study is to determine the pharmacokinetics (PKs) of EACA in infants aged 6-24 months undergoing craniofacial reconstruction surgery. Cohorts of six infants were enrolled sequentially to one of the three escalating loading dose-continuous i.v. infusion (CIVI) regimens: 25 mg kg(-1), 10 mg kg(-1) h(-1); 50 mg kg(-1), 20 mg kg(-1) h(-1); 100 mg kg(-1), 40 mg kg(-1) h(-1). Plasma EACA concentrations were determined using a validated high-performance liquid chromatography-tandem mass spectrometry assay. A population non-linear mixed effects modelling approach was used to characterize EACA PKs. Population PK parameters of EACA were estimated using a two-compartment disposition model with weight expressed as an allometric covariate and an age effect. The typical patient in this study had an age of 38.71 weeks and a weight of 8.82 kg. PK parameters for this typical patient were: pre-/postoperative plasma drug clearance of 32 ml min(-1) (3.6 ml kg(-1) min(-1)), inter-compartmental clearance of 42.4 ml min(-1) (4.8 ml min(-1) kg(-1)), central volume of distribution of 1.27 litre (0.14 litre kg(-1)), and peripheral volume of distribution of 2.53 litre (0.29 litre kg(-1)). Intra-operative clearance and central volume of distribution were 89% and 80% of the pre-/postoperative value, respectively. EACA clearance increased with weight and age. The dependence of clearance on body weight supports weight-based dosing. Based on this study, a loading dose of 100 mg kg(-1) followed by a CIVI of 40 mg kg(-1) h(-1) is appropriate to maintain target plasma EACA concentrations in children aged 6-24 months undergoing these procedures.
Product Description
Supelco
Aminocaproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)
Sigma-Aldrich
6-Aminocaproic acid, BioUltra, ≥99%
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Sigma-Aldrich
6-Aminocaproic acid, SAJ special grade, ≥99.0%