Merck
  • A bis(triazole)benzamide receptor for the complexation of halide anions and neutral carboxylic acid guests. Guest-controlled topicity and self-assembly.
A bis(triazole)benzamide receptor for the complexation of halide anions and neutral carboxylic acid guests. Guest-controlled topicity and self-assembly.
Organic & biomolecular chemistry (2012-12-18)
Fátima García, Juan Aragó, Rafael Viruela, Enrique Ortí, Luis Sánchez
ABSTRACT
Bis(triazole)benzamide 1 has been readily synthesized by means of Cu-catalyzed 1,3-dipolar cycloaddition and its ability to bind halide anions and neutral gallic acid derivative 12GA has been theoretically and experimentally investigated. The cavity defined by the N-H amide group and the vicinal aromatic hydrogens is suitable to form H-bonding arrays with halide guests. The stability of complexes 1·Cl(-) and 1·Br(-) is very similar, as DFT calculations predict and (1)H NMR titration experiments confirm. The zigzag "anti" conformation of the molecule generates two regions with complementary positive and negative potentials that favor the statistical complexation of two molecules of the neutral carboxylic acid 12GA. This guest-controlled topicity demonstrates the versatility of this class of receptor to bind species of different nature. The amide group determines the complexation of both anionic and neutral species by primary acid-base interactions.
Sigma-Aldrich
Bromine, SAJ first grade, ≥97.0%
Sigma-Aldrich
Bromine, JIS special grade, ≥99.0%
Sigma-Aldrich
Bromine, ≥99.99% trace metals basis
Sigma-Aldrich
Bromine, ACS reagent, ≥99.5%
Sigma-Aldrich
Bromine, reagent grade
Sigma-Aldrich
Bromine water, CP