Merck
Evolutionary genetics: no coming back from neverland.
Current biology : CB (2012-12-12)
Artyom Kopp
ABSTRACT
Host-plant specialization plays a key role in insect evolution, but little is known about its molecular basis. A new paper shows that a cactus-feeding fly became restricted to its host by changes in an enzyme that converts dietary sterols into essential hormones.
Sigma-Aldrich
5α-Cholest-7-en-3β-ol