Merck
  • A new drug delivery system for intravenous coronary thrombolysis with thrombus targeting and stealth activity recoverable by ultrasound.
A new drug delivery system for intravenous coronary thrombolysis with thrombus targeting and stealth activity recoverable by ultrasound.
Journal of the American College of Cardiology (2012-11-20)
Hiroyuki Kawata, Yoshiko Uesugi, Tsunenari Soeda, Yasuhiro Takemoto, Ji-Hee Sung, Kiyotaka Umaki, Keiji Kato, Kenichi Ogiwara, Keiji Nogami, Kenichi Ishigami, Manabu Horii, Shiro Uemura, Midori Shima, Yasuhiko Tabata, Yoshihiko Saito
ABSTRACT
The purpose of this study was to develop a new intelligent drug delivery system for intracoronary thrombolysis with a strong thrombolytic effect without increasing bleeding risk. Rapid recanalization of an occluded coronary artery is essential for better outcomes in acute myocardial infarction. Catheter-based recanalization is widely accepted, but it takes time to transport patients. Although the current fibrinolytic therapy can be started quickly, it cannot achieve a high reperfusion rate. Recently, we generated nanoparticles comprising tissue-type plasminogen activator (tPA), basic gelatin, and zinc ions, which suppress tPA activity by 50% with 100% recovery by ultrasound (US) in vitro. The thrombus-targeting property of nanoparticles was examined by an in vitro binding assay with von Wilbrand factor and with a mouse arterial thrombosis model in vivo. The thrombolytic efficacy of nanoparticles was evaluated with a swine acute myocardial infarction model. Nanoparticles bound to von Wilbrand factor in vitro and preferentially accumulated at the site of thrombus in a mouse model. In a swine acute myocardial infarction model, plasma tPA activity after intravenous injection of nanoparticles was approximately 25% of tPA alone and was recovered completely by transthoracic US (1.0 MHz, 1.0 W/cm(2)). During US application, plasma tPA activity near the affected coronary artery was recovered and was higher than that near the femoral artery. Although treatment with tPA alone (55,000 IU/kg) recanalized the occluded coronary artery in only 1 of 10 swine, nanoparticles containing the same dose of tPA with US achieved recanalization in 9 of 10 swine within 30 min. We developed an intelligent drug delivery system with promising potential for better intravenous coronary thrombolysis.
Sigma-Aldrich
Zinc acetate dihydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc acetate dihydrate, reagent grade
Sigma-Aldrich
Zinc acetate dihydrate, puriss., E 650, 99-102%
Sigma-Aldrich
Zinc acetate dihydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Zinc acetate dihydrate, 99.999% trace metals basis
Sigma-Aldrich
Zinc acetate dihydrate, ACS reagent, ≥98%