Merck
  • Arrhythmia associated with tetracaine in an extremely low birth weight premature infant.
Arrhythmia associated with tetracaine in an extremely low birth weight premature infant.
Pediatrics (2012-11-07)
Halima Maulidi, Carol McNair, Neil Seller, Joel Kirsh, Timothy J Bradley, Steven C Greenway, Chris Tomlinson
ABSTRACT
Infants in NICUs undergo a variety of painful procedures. The management of pain has become an integral part of newborn infant care with the use of both systemic and topical agents to provide analgesia and anesthesia for procedural pain. Tetracaine and prilocaine-lidocaine are the 2 topical anesthetics most frequently used. Tetracaine belongs to an ester group of local anesthetics available as a topical 4% gel (Ametop, Smith and Nephew, Canada). The major side effects reported when using topical anesthetics are cutaneous reactions. There are no definite reports of systemic toxicity in the published literature. We present a recent case of an extremely low birth weight premature infant who developed a clinically significant arrhythmia after topical tetracaine was applied before the insertion of a peripherally inserted central catheter. The infant had no other identifiable cause for the resulting bradycardia that occurred only after Ametop was applied. The cardiac symptoms resolved with treatment. This case highlights a significant potential adverse event when using topical tetracaine.
Sigma-Aldrich
Tetracaine, ≥98% (TLC)
Supelco
Tetracaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetracaine hydrochloride, ≥99%
Tetracaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Tetracaine for system suitability, European Pharmacopoeia (EP) Reference Standard