Merck
  • Growth and spectroscopic investigation of a new crystal for NLO applications: C₁₀H₂ ₀KN₅O₉.
Growth and spectroscopic investigation of a new crystal for NLO applications: C₁₀H₂ ₀KN₅O₉.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-10-16)
Redrothu Hanumantharao, S Kalainathan
ABSTRACT
Nonlinear optical crystals of Bis (l-glutamine) potassium nitrate (BGPN) were grown by slow evaporation technique at ambient temperature. Solubility and metastable zone width of BGPN in aqueous solution were determined. The grown crystal was characterized by single crystal XRD, Powder XRD, FT-IR (1)H NMR, EDAX, mass and optical spectroscopic techniques. Single crystal XRD revealed that compound crystallizes in orthorhombic system with non-centrosymmetric space group P2(1)2(1)2(1). The sharp peaks from powder XRD spectrum show the high crystallinity of the grown crystal. FT-IR confirms the presence of functional groups and molecular structure was confirmed by (1)H NMR spectrum of the grown crystal. Molecular mass of BGPN sample has been verified by high resolution mass spectroscopic analysis. The presence of potassium in the compound and composition of grown crystals was confirmed on the basis of energy dispersive analysis of X-ray (EDAX). Thermal stability of the grown crystal was studied by TGA-DTA analysis. An optical UV-Vis-NIR spectrum for BGPN sample was recorded in the range of 190-1100 nm. Fluorescence studies shows material BGPN emits blue fluorescence. Second harmonic generation (SHG) studies have been performed by famous Kurtz powder technique with reference to standard potassium dihydrogen phosphate single crystals (KDP). It is found from this technique that SHG efficiency of BGPN is in comparison to that of standard KDP crystals.
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium nitrate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Supelco
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)